Boron carbide ≥99.4%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40504-18
40504-30
CAAA40504-18EA
88.48
CAD
CAAA40504-18
CAAA40504-30
Boron carbide ≥99.4%
Boron carbide
MDL: MFCD00011520
CAS No.: 12069-32-8
Molecular Formula: B4C
Formula Weight: 55.26
Physical Form: 1-7 Micron Powder
Density: 2.52
Odor: Odorless
Merck Reference: 14,1344
MDL No.: MFCD00011520
Formula:
CB₄ MW: 55.25 Boiling Pt: ∼3500 °C (1013 hPa) Melting Pt: 2350 Density: 2.52 Storage Temperature: Ambient |
MDL Number:
MFCD00011520 CAS Number: 12069-32-8 EINECS: 235-111-5 Merck Index: 14,1344 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...