Tungsten(IV) carbide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40502-18
40502-30
CAAA40502-18EA
98.59
CAD
CAAA40502-18
CAAA40502-30
Tungsten(IV) carbide ≥99% (metals basis)
Tungsten(IV) carbide
MDL: MFCD00011464
Purity: 99%
CAS No.: 12070-12-1
Molecular Formula: WC
Formula Weight: 195.86
Physical Form: -100+270 Mesh Powder
Density: 15.63
Melting Point: 2870°
Merck Reference: 14,9815
UN No.: 3178
MDL No.: MFCD00011464
Formula:
CW Boiling Pt: 6000 °C (1013 hPa) Melting Pt: 2870 °C Density: 15.63 Storage Temperature: Ambient |
MDL Number:
MFCD00011464 CAS Number: 12070-12-1 EINECS: 235-123-0 UN: 3178 ADR: 4.1,III Merck Index: 14,9815 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...