Nickel (II) sulfamate 50% (w/w) in aqueous solution, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickel (II) sulphamidate, Nickel bis(sulphamidate), Nickel (II) sulfamidate, Nickel sulfamate, Nickel bis(sulfamidate), Nickel sulphamate
40501-K2
40501-8M
CAAA40501-K2EA
97.96
CAD
CAAA40501-K2
CAAAA40501-8M
Nickel (II) sulfamate 50% (w/w) in aqueous solution, Reagent Grade
Nickel (II) sulphamate
Nickel plating
CAS No.: 13770-89-3
Molecular Formula: H4N2NiO6S2
Formula Weight: 250.86(anhy)
Physical Form: Liquid
Appearance: Green
Odor: Odorless
UN No.: 3082
MDL No.: MFCD00137261
Nickel plating
Formula:
H₄N₂NiO₆S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00137261 CAS Number: 13770-89-3 EINECS: 237-396-1 UN: 3082 ADR: 9,III |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...