Iron ≥99.99% (metals basis), rods, Ø 12 mm (0.47 in)

Supplier: Thermo Scientific Chemicals

40499-DJ 40499-HF
CAAA40499-DJEA 856.36 CAD
CAAA40499-DJ CAAA40499-HF
Iron ≥99.99% (metals basis), rods, Ø 12 mm (0.47 in)
Iron

MDL: MFCD00010999


Purity: 99.99% CAS No.: 7439-89-6 Physical Form: ≈0.89g/mm MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR