Iron ≥99.99% (metals basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

40493-FF 40493-FI 40493-GH
CAAA40493-FFEA 82.95 CAD
CAAA40493-FF CAAA40493-FI CAAA40493-GH
Iron ≥99.99% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Iron

MDL: MFCD00010999


Purity: 99.99% CAS No.: 7439-89-6 Physical Form: ≈0.49g/25x25mm MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR