Niobium ≥99.8% (metals basis), powder APS 1-5 µm

Supplier: Thermo Scientific Chemicals
Danger

40478-09 40478-22 40478-30
CAAA40478-09EA 82.63 CAD
CAAA40478-09 CAAA40478-22 CAAA40478-30
Niobium ≥99.8% (metals basis), powder APS 1-5 µm
Niobium

Used in high temperature and super alloys of Zn and Ti


Purity: 99.8% CAS No.: 7440-03-1 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011126


Used in high temperature and super alloys of Zn and Ti

Formula: Nb
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR