tert-Butyl methyl ether 99+% HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

40477-K2 40477-K7 40477-M1 40477-M6
CAAA40477-K2EA 161.16 CAD
CAAA40477-K2 CAAA40477-K7 CAAA40477-M1 CAAA40477-M6
tert-Butyl methyl ether 99+% HPLC
tert-Butyl methyl ether

Chromatography solvent


Purity: 99+% CAS No.: 1634-04-4 Molecular Formula: (CH3)3COCH3 Formula Weight: 88.15 Flash Point: -25°(-13°F) Physical Form: Liquid Density: 0.742 Refractive Index: 1.3690 Odor: Mild Melting Point: -109° Boiling Point: 55-56° Beilstein Reference: 1730942 Merck Reference: 14,6032 UN No.: 2350 MDL No.: MFCD00008812


Chromatography solvent

Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2
Melting Pt: -108.6
Density: 0.742
Flash Pt: –25 °C (–13 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2350
ADR: 3 II
Merck Index: 14,6032
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR