Bismuth(III) nitrate basic Bi₂O₃ 79%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
Ceramic glazes and enamel fluxes
CAS No.: 1304-85-4
Molecular Formula: Bi5O(OH)9(NO3)4 (approximate)
Formula Weight: 1461.98
Physical Form: Powder
Density: 6.04
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 260° dec.
Solubility: Practically insoluble in water and alcohol. Soluble in dilute HNO3 and HCl
Merck Reference: 14,1284
UN No.: 1477
MDL No.: MFCD00064844
Ceramic glazes and enamel fluxes
Formula:
Bi₅O(OH)₉(NO₃)₄ MW: 1461.99 g/mol Melting Pt: 260 °C (dec.) Density: 6.04 Storage Temperature: Ambient |
MDL Number:
MFCD00064844 CAS Number: 1304-85-4 EINECS: 215-136-8 UN: 1477 ADR: 5.1,II Merck Index: 14,1284 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...