Dirhenium heptaoxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
As a catalyst for oxidation/reduction reactions
Purity: 99.9%
CAS No.: 1314-68-7
Molecular Formula: O7Re2
Formula Weight: 484.40
Physical Form: Crystalline, Ampouled under argon
Density: 6.103
Appearance: Yellow to pale green
Odor: Odorless
Melting Point: 297°
Boiling Point: 360°
Solubility: Dissolves in water to form HReO4. Very soluble in alcohol. Soluble in alkalies and acids
Merck Reference: 14,8178
UN No.: 3085
MDL No.: MFCD00011199
As a catalyst for oxidation/reduction reactions
Formula:
O₇Re₂ Boiling Pt: 360 °C Melting Pt: 297 °C Density: 6.103 Storage Temperature: Ambient |
MDL Number:
MFCD00011199 CAS Number: 1314-68-7 EINECS: 215-241-9 UN: 3085 ADR: 5.1,III Merck Index: 14,8178 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...