Titanium(IV) oxide ≥99.9% (metals basis), REacton®, high surface area

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

40458-04 40458-09
CAAA40458-04EA 156.58 CAD
CAAA40458-04 CAAA40458-09
Titanium(IV) oxide ≥99.9% (metals basis), REacton®, high surface area
Titanium (IV) oxide

Photocatalyst, catalyst, catalyst support


Purity: 99.9% CAS No.: 13463-67-7 Molecular Formula: O2Ti Formula Weight: 79.90 Physical Form: -325 Mesh Powder, S.A. 180-300 m2/g Density: 4.26 Appearance: White Odor: Odorless Melting Point: 1830-1850° Boiling Point: 2500-3000° Solubility: Insoluble in water, HCl, HNO3, dilute H2SO4; soluble in hot, conc. H2SO4, HF, and alkalies Merck Reference: 14,9472 MDL No.: MFCD00011269


Photocatalyst, catalyst, catalyst support

Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2900
Melting Pt: 1855
Density: 4.26
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 14,9472

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR