Nickel chromium alloy (80:20 w%) ≥99% (metals basis), wire, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40438-G9
40438-H4
40438-H9
CAAA40438-G9EA
89.27
CAD
CAAA40438-G9
CAAA40438-H4
CAAA40438-H9
Nickel chromium alloy (80:20 w%) ≥99% (metals basis), wire, Ø 1.0 mm (0.04 in)
Nickel Chromium alloy (80:20 w%)
MDL: MFCD00673110
Purity: 99%
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈6.6g/m
Odor: Odorless
Storage Temperature:
Ambient |
MDL Number:
MFCD00673110 CAS Number: 11106-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...