Gold(I) chloride ≥99.99% (metals basis) Au ≥ 84.2%, Premion®
Supplier: Thermo Scientific Chemicals
40432-01
40432-03
40432-06
CAAA40432-01EA
143.46
CAD
CAAA40432-01
CAAA40432-03
CAAA40432-06
Gold(I) chloride ≥99.99% (metals basis) Au ≥ 84.2%, Premion®
Gold (I) chloride
MDL: MFCD00046175
Notes: Decomposes in water
Purity: 99.99%
CAS No.: 10294-29-8
Molecular Formula: AuCl
Formula Weight: 232.42
Physical Form: Powder
Density: 7.57
Melting Point: 289° dec.
Merck Reference: 14,4515
UN No.: 3260
MDL No.: MFCD00046175
Formula:
AuCl Melting Pt: 289 °C (dec.) Density: 7.57 |
MDL Number:
MFCD00046175 CAS Number: 10294-29-8 EINECS: 233-655-8 UN: 3260 ADR: 8,III Merck Index: 14,4515 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...