Aluminium ≥99.99% (metals basis), slug, Length: 8.0 mm (0.315 in), Ø 1.98 mm (0.078 in)

Supplier: Thermo Scientific Chemicals

40417-09 40417-18 40417-30
CAAA40417-09EA 90.38 CAD
CAAA40417-09 CAAA40417-18 CAAA40417-30
Aluminium ≥99.99% (metals basis), slug, Length: 8.0 mm (0.315 in), Ø 1.98 mm (0.078 in)
Aluminium

Evaporation or metallizing source material


Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈140pc/10g Odor: Odorless MDL No.: MFCD00134029


Evaporation or metallizing source material

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR