Zinc bromide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40413-18
40413-30
40413-A1
CAAA40413-18EA
81.84
CAD
CAAA40413-18
CAAA40413-30
CAAA40413-A1
Zinc bromide ≥99.9% (metals basis)
Zinc bromide
MDL: MFCD00011294
Fieser: 2,463 13,349 15,368 16,389 18,409 19,409 20,438 21,493
Purity: 99.9%
CAS No.: 7699-45-8
Molecular Formula: ZnBr2
Formula Weight: 225.19
Physical Form: Crystalline
Density: 4.201
Refractive Index: 1.5452
Sensitivity: Hygroscopic
Appearance: White to pale cream
Melting Point: 394°
Boiling Point: 650°
Merck Reference: 14,10129
UN No.: 3260
MDL No.: MFCD00011294
Formula:
Br₂Zn Boiling Pt: 650 °C Melting Pt: 394 °C Density: 4.201 Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS Number: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,II Merck Index: 14,10129 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...