Disodium tetrabromopalladate ≥99.95% (metals basis) Pd ≥ 22.0%, Premion®

Supplier: Thermo Scientific Chemicals
Warning

40407-03 40407-06
CAAA40407-03EA 161.16 CAD
CAAA40407-03 CAAA40407-06 CAAA40407-09
Disodium tetrabromopalladate ≥99.95% (metals basis) Pd ≥ 22.0%, Premion®
Disodium tetrabromopalladate

MDL: MFCD00046199


Purity: 99.95% CAS No.: 50495-13-1 Molecular Formula: Na2PdBr4 Formula Weight: 472.02 Physical Form: Powder

Formula: Br₄Na₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00046199
CAS Number: 50495-13-1
EINECS: 256-605-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR