Platinium(IV) oxide, anhydrous ≥99.95% (metals basis) Pt ≥ 84.4%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Platinum dioxide, Adam's type catalyst
40402-01
40402-03
40402-06
CAAA40402-01EA
135.41
CAD
CAAA40402-01
CAAA40402-03
CAAA40402-06
Platinium(IV) oxide, anhydrous ≥99.95% (metals basis) Pt ≥ 84.4%, Premion®
Platinium (IV) oxide
MDL: MFCD00011184
Purity: 99.95%
CAS No.: 1314-15-4
Molecular Formula: O2Pt
Formula Weight: 227.09
Physical Form: Powder
Density: 10.2
Appearance: Dark brown
Odor: Odorless
Melting Point: 450°
Merck Reference: 14,7527
UN No.: 1479
MDL No.: MFCD00011184
Formula:
O₂Pt MW: 227.08 g/mol Melting Pt: 450 °C Density: 10.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011184 CAS Number: 1314-15-4 EINECS: 215-223-0 UN: 1479 ADR: 5.1,II Merck Index: 14,7527 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...