Bismuth chloride oxide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth oxychloride, Bismuth (III) oxychloride, Bismuth(III) chloride oxide

40396-18 40396-30 40396-A1
CAAA40396-18EA 90.38 CAD
CAAA40396-18 CAAA40396-30 CAAA40396-A1
Bismuth chloride oxide 98%
Bismuth chloride oxide

Used as pigment, in manufacture of artificial pearls, dry-cell cathodes


Purity: 98% CAS No.: 7787-59-9 Molecular Formula: BiClO Formula Weight: 260.43 Physical Form: Powder, typically 2 micron max Density: 7.72 Refractive Index: 2.15 Appearance: Very pale brown Odor: Odorless Melting Point: dec. Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid Merck Reference: 14,1263 MDL No.: MFCD00010895


Used as pigment, in manufacture of artificial pearls, dry-cell cathodes

Formula: BiClO
MW: 260.43 g/mol
Melting Pt: Dec.
Density: 7.72
Storage Temperature: Ambient
MDL Number: MFCD00010895
CAS Number: 7787-59-9
EINECS: 232-122-7
Merck Index: 14,1263

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR