Bismuth(III) oxide 99.975% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

40394-18 40394-30 40394-A1
CAAA40394-18EA 119.92 CAD
CAAA40394-18 CAAA40394-30 CAAA40394-A1
Bismuth(III) oxide 99.975% (metals basis)
Bismuth (III) oxide

MDL: MFCD00003462


Purity: 99.975% CAS No.: 1304-76-3 Molecular Formula: Bi2O3 Formula Weight: 465.96 Physical Form: Powder, typically 4 micron Density: 8.9 Appearance: Yellow Melting Point: 817° Boiling Point: ca 1890° Merck Reference: 14,1273 MDL No.: MFCD00003462

Formula: BiO₃
Boiling Pt: ∼1890 °C
Melting Pt: 817 °C
Density: 8.9
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 14,1273

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR