Copper ≥99.9% (metals basis), foil, annealed, Thickness: 0.675 mm (0.027 in)

Supplier: Thermo Scientific Chemicals

40374-CU 40374-CV
CAAA40374-CUEA 199.08 CAD
CAAA40374-CU CAAA40374-CV
Copper ≥99.9% (metals basis), foil, annealed, Thickness: 0.675 mm (0.027 in)
Copper

MDL: MFCD00010965


Purity: 99.9% CAS No.: 7440-50-8 Physical Form: ≈241g/20x20cm, Temper: soft Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR