Cerium tribromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40373-14
40373-03
40373-06
CAAAA40373-14EA
1412.52
CAD
CAAAA40373-14
CAAA40373-03
CAAA40373-06
Cerium tribromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Cerium tribromide
MDL: MFCD00016004
Purity: 99.9%
CAS No.: 14457-87-5
Molecular Formula: CeBr3
Formula Weight: 379.83
Physical Form: Powder, Ampouled under argon
Density: 5.18
Appearance: White to pale grey
Odor: Odorless
Melting Point: 733°
Boiling Point: 1560°
Merck Reference: 14,1995
MDL No.: MFCD00016004
Formula:
CeBr₃ Boiling Pt: 1560 °C Melting Pt: 733 °C Density: 5.18 Storage Temperature: Ambient |
MDL Number:
MFCD00016004 CAS Number: 14457-87-5 EINECS: 238-447-0 Merck Index: 14,1995 |
Specification Test Results
Form | Powder, Ampouled under argon |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...