Antimony (III) sulfide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40371-09
40371-18
40371-30
CAAA40371-09EA
139.04
CAD
CAAA40371-09
CAAA40371-18
CAAA40371-30
Antimony (III) sulfide ≥99.9% (metals basis)
Antimony (III) sulphide
Vacuum deposition
Purity: 99.9%
CAS No.: 1345-04-6
Molecular Formula: Sb2S3
Formula Weight: 339.68
Physical Form: 3-12mm Pieces
Melting Point: 550°
UN No.: 1549
MDL No.: MFCD00011217
Vacuum deposition
Formula:
S₃Sb₂ Boiling Pt: 1150 °C (1013 hPa) Melting Pt: 550 °C Density: 4.12 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011217 CAS Number: 1345-04-6 EINECS: 215-713-4 UN: 1549 ADR: 6.1,III Merck Index: 14,713 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...