Arsenic(III) oxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40370-09
40370-18
40370-30
CAAA40370-09EA
95.12
CAD
CAAA40370-09
CAAA40370-18
CAAA40370-30
Arsenic(III) oxide ≥99.99% (metals basis)
Arsenic (III) oxide
MDL: MFCD00003433
Purity: 99.99%
CAS No.: 1327-53-3
Molecular Formula: As2O3
Formula Weight: 197.84
Physical Form: -100 Mesh Powder
Density: 3.74
Melting Point: 312.3°
Boiling Point: 465° subl.
Merck Reference: 14,804
UN No.: 1561
MDL No.: MFCD00003433
Formula:
As₂O₃ Boiling Pt: 465 °C (subl.) Melting Pt: 312.3 °C Density: 3.74 Storage Temperature: Ambient |
MDL Number:
MFCD00003433 CAS Number: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1,II Merck Index: 14,804 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...