Tungsten dioxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

40367-06 40367-14 40367-22
CAAA40367-06EA 125.61 CAD
CAAA40367-06 CAAA40367-14 CAAA40367-22
Tungsten dioxide ≥99.9% (metals basis)
Tungsten dioxide

MDL: MFCD00049709 Insoluble in water. Soluble in acid, KOH


Purity: 99.9% CAS No.: 12036-22-5 Molecular Formula: WO2 Formula Weight: 215.85 Physical Form: -100 Mesh Powder Density: 12.11 Melting Point: 1500-1600° (in N2) Solubility: Insoluble in water. Soluble in acid, KOH MDL No.: MFCD00049709

Formula: O₂W
MW: 215.84
Melting Pt: 1500…1600 °C (in N₂)
Density: 10.8
Storage Temperature: Ambient
MDL Number: MFCD00049709
CAS Number: 12036-22-5
EINECS: 234-842-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR