Thallium(I) sulfate ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40342-09
40342-18
40342-30
CAAA40342-09EA
113.6
CAD
CAAA40342-09
CAAA40342-18
CAAA40342-30
Thallium(I) sulfate ≥99.5% (metals basis)
Dithallium sulphate
MDL: MFCD00011278
Solubility in water increases with temperature
Purity: 99.5% min
CAS No.: 7446-18-6
Molecular Formula: Tl2SO4
Formula Weight: 504.80
Physical Form: Crystalline
Density: 6.77
Melting Point: 632°
Boiling Point: dec.
Solubility: Solubility in water increases with temperature
Merck Reference: 14,9269
UN No.: 1707
MDL No.: MFCD00011278
Formula:
O₄STl₂ Boiling Pt: Dec. Melting Pt: 632 °C Density: 6.77 Storage Temperature: Ambient |
MDL Number:
MFCD00011278 CAS Number: 7446-18-6 EINECS: 231-201-3 UN: 1707 ADR: 6.1,II Merck Index: 14,9269 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...