Indium ≥99.99% (metals basis), spheres, Ø 1 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

40339-06 40339-18 40339-30
CAAA40339-06EA 131.14 CAD
CAAA40339-06 CAAA40339-18 CAAA40339-30
Indium ≥99.99% (metals basis), spheres, Ø 1 mm (0.04 in)
Indium

MDL: MFCD00134048


Purity: 99.99% CAS No.: 7440-74-6 Physical Form: Tolerance +/- 0.1mm Odor: Odorless MDL No.: MFCD00134048

Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR