Indium ≥99.999% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.12 in)

Supplier: Thermo Scientific Chemicals

40338-09 40338-18 40338-30
CAAA40338-09EA 208.56 CAD
CAAA40338-09 CAAA40338-18 CAAA40338-30
Indium ≥99.999% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.12 in)
Indium

MDL: MFCD00134048


Purity: 99.999% CAS No.: 7440-74-6 Physical Form: Spherical shot Appearance: Silver Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR