Ruthenium(IV) oxide ≥99.95% (metals basis) Ru typ. 74%, Premion®, Electronic Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
40336-04
40336-09
CAAA40336-04EA
402.9
CAD
CAAA40336-04
CAAA40336-09
Ruthenium(IV) oxide ≥99.95% (metals basis) Ru typ. 74%, Premion®, Electronic Grade
Ruthenium (IV) oxide
Semiconductor industry for the manufacture of resistor pastes
Purity: 99.95%
CAS No.: 12036-10-1
Molecular Formula: RuO2
Formula Weight: 133.07
Physical Form: Sub-micron Powder, Surface Area 45-65m2/g
Density: 6.97
Appearance: Black
Odor: Odorless
Melting Point: 1200° subl.
UN No.: 1479
MDL No.: MFCD00011210
Semiconductor industry for the manufacture of resistor pastes
Formula:
O₂Ru Melting Pt: 1200 °C (subl.) Density: 6.97 Storage Temperature: Ambient |
MDL Number:
MFCD00011210 CAS Number: 12036-10-1 EINECS: 234-840-6 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...