Gold 14kt (Au ≥ 58.3%), yellow, wire, Ø 1.63 mm (0.064 in)

Supplier: Thermo Scientific Chemicals

40329-BS 40329-BU
CAAA40329-BSEA 748.92 CAD
CAAA40329-BS CAAA40329-BU CAAA40329-BW
Gold 14kt (Au ≥ 58.3%), yellow, wire, Ø 1.63 mm (0.064 in)
Gold

MDL: MFCD00003436


Molecular Formula: % Concentration CuZnAg Physical Form: ≈28g/m Appearance: Yellow MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR