Lanthanum hexaboride ≥99.5% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
40325-09
40325-18
40325-30
CAAA40325-09EA
112.34
CAD
CAAA40325-09
CAAA40325-18
CAAA40325-30
Lanthanum hexaboride ≥99.5% (REO, rare earth oxide basis)
Lanthanum hexaboride
MDL: MFCD00151350
Insoluble in water, HCl
Purity: 99.5%
CAS No.: 12008-21-8
Molecular Formula: LaB6
Formula Weight: 203.78
Physical Form: -325 Mesh Powder
Density: 2.61
Melting Point: 2210°
Solubility: Insoluble in water, HCl
MDL No.: MFCD00151350
Formula:
B₆La Melting Pt: 2210 °C Density: 2.61 Storage Temperature: Ambient |
MDL Number:
MFCD00151350 CAS Number: 12008-21-8 EINECS: 234-531-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...