Cobalt(II) bromide, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cobaltous bromide
40319-03
40319-06
CAAA40319-03EA
175.38
CAD
CAAA40319-03
CAAA40319-06
Cobalt(II) bromide, ultra dry ≥99.99% (metals basis)
Cobalt (II) bromide
MDL: MFCD00016017
Purity: 99.99%
CAS No.: 7789-43-7
Molecular Formula: CoBr2
Formula Weight: 218.75
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.909
Appearance: Green
Melting Point: 678° (in N2)
Merck Reference: 14,2435
MDL No.: MFCD00016017
Formula:
Br₂Co MW: 218.74 g/mol Melting Pt: 678 °C (in N₂) Density: 4.909 Storage Temperature: Ambient |
MDL Number:
MFCD00016017 CAS Number: 7789-43-7 EINECS: 232-166-7 Merck Index: 14,2435 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...