Yttrium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

40299-DH 40299-DJ 40299-DM 40299-HF
CAAA40299-DHEA 371.3 CAD
CAAA40299-DH CAAA40299-DJ CAAA40299-DM CAAA40299-HF
Yttrium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Yttrium

MDL: MFCD00011468


Purity: 99.9% CAS No.: 7440-65-5 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.14g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Y
Boiling Pt: 2927 °C (1013 hPa)
Melting Pt: 1525 °C
Density: 4.472 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR