Samarium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

40298-DH 40298-DJ 40298-DM
CAAA40298-DHEA 418.7 CAD
CAAA40298-DH CAAA40298-DJ CAAA40298-DM CAAA40298-HF
Samarium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Samarium

MDL: MFCD00011233


Purity: 99.9% CAS No.: 7440-19-9 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.96g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Sm
Boiling Pt: 1900 °C (1013 hPa)
Melting Pt: 1076.85 °C
Density: 7.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR