Samarium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

40297-DH 40297-DJ 40297-DM 40297-HF
CAAA40297-DHEA 271.76 CAD
CAAA40297-DH CAAA40297-DJ CAAA40297-DM CAAA40297-HF
Samarium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Samarium

MDL: MFCD00011233


Purity: 99.9% CAS No.: 7440-19-9 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.24g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1900
Melting Pt: 1076.85
Density: 7.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR