Praseodymium ≥99.1% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
40295-DH
40295-DJ
40295-DM
40295-HF
CAAA40295-DHEA
364.98
CAD
CAAA40295-DH
CAAA40295-DJ
CAAA40295-DM
CAAA40295-HF
Praseodymium ≥99.1% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Praseodymium
MDL: MFCD00011174
Purity: 99.1%
CAS No.: 7440-10-0
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.22g/mm, Cast rod, diameter is approximate
Odor: Odorless
Formula:
Pr MW: 140.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011174 CAS Number: 7440-10-0 EINECS: 231-120-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...