Neodymium ≥99.5% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

40294-DH 40294-DJ 40294-DM 40294-HF
CAAA40294-DHEA 347.6 CAD
CAAA40294-DH CAAA40294-DJ CAAA40294-DM CAAA40294-HF
Neodymium ≥99.5% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Neodymium

MDL: MFCD00011130


Purity: 99.5% CAS No.: 7440-00-8 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.89g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Nd
MW: 144.24
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR