Lanthanum ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals
Danger

40293-DH 40293-DJ 40293-DM 40293-HF
CAAA40293-DHEA 316 CAD
CAAA40293-DH CAAA40293-DJ CAAA40293-DM CAAA40293-HF
Lanthanum ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Lanthanum

MDL: MFCD00011066


Purity: 99.9% CAS No.: 7439-91-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.80g/mm, Cast rod, diameter is approximate Appearance: Silver-gray UN No.: 3208 MDL No.: MFCD00011066

Formula: La
Boiling Pt: 3464 °C (1013 hPa)
Melting Pt: 920 °C
Density: ∼6,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR