Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Scientific Chemicals
40290-DH
40290-HF
CAAA40290-DHEA
1112.32
CAD
CAAA40290-DH
CAAA40290-HF
Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Gadolinium
MDL: MFCD00011022
Purity: 99.9%
CAS No.: 7440-54-2
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.97g/mm, Cast rod, diameter is approximate
MDL No.: MFCD00011022
Formula:
Gd Storage Temperature: Ambient |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...