Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

40288-DH 40288-DJ 40288-HF
CAAA40288-DHEA 1124.96 CAD
CAAA40288-DH CAAA40288-DJ CAAA40288-HF
Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Erbium

MDL: MFCD00010987


Purity: 99.9% CAS No.: 7440-52-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈1.2g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Er
MW: 167.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR