Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

40287-DH 40287-DJ 40287-HF
CAAA40287-DHEA 443.98 CAD
CAAA40287-DH CAAA40287-DJ CAAA40287-HF
Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Erbium

MDL: MFCD00010987


Purity: 99.9% CAS No.: 7440-52-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.29g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Er
MW: 167.26
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR