Dysprosium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Scientific Chemicals
40286-DH
40286-DJ
40286-DM
40286-HF
CAAA40286-DHEA
1019.1
CAD
CAAA40286-DH
CAAA40286-DJ
CAAA40286-DM
CAAA40286-HF
Dysprosium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Dysprosium
MDL: MFCD00010982
Purity: 99.9%
CAS No.: 7429-91-6
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈1.1g/mm, Cast rod, diameter is approximate
Odor: Odorless
Formula:
Dy MW: 162.5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...