Dysprosium ≥99.7% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

40285-DH 40285-DJ 40285-DM 40285-HF
CAAA40285-DHEA 404.48 CAD
CAAA40285-DH CAAA40285-DJ CAAA40285-DM CAAA40285-HF
Dysprosium ≥99.7% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Dysprosium

MDL: MFCD00010982


Purity: 99.7% CAS No.: 7429-91-6 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.27g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Dy
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR