Cerium ≥99.8% (metals basis excluding Ta), rods Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals
Danger

40284-DH 40284-DJ 40284-DM
CAAA40284-DHEA 439.24 CAD
CAAA40284-DH CAAA40284-DJ CAAA40284-DM
Cerium ≥99.8% (metals basis excluding Ta), rods Ø 12.7 mm (0.5 in)
Cerium

Purity: 99.8% CAS No.: 7440-45-1 Packaged in mylar under argon Physical Form: ≈0.86g/mm, Cast rod, diameter is approximate Appearance: Silver Odor: Odorless UN No.: 1333 MDL No.: MFCD00010924

Formula: Ce
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 795 °C
Density: ∼6,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR