Magnesium oxide substrate 10x10x0.5mm, polished one side 110 orientation

Supplier: Thermo Scientific Chemicals

40272-KT 40272-KU
CAAA40272-KTEA 161.16 CAD
CAAA40272-KT CAAA40272-KU
Magnesium oxide substrate 10x10x0.5mm, polished one side 110 orientation
Magnesium oxide

MDL: MFCD00011109


Molecular Formula: MgO MDL No.: MFCD00011109

Formula: MgO
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: 2800 °C
Density: 3.58 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR