Hafnium dioxide, tech.

Supplier: Thermo Scientific Chemicals

40270-22 40270-36
CAAA40270-22EA 387.1 CAD
CAAA40270-22 CAAA40270-36
Hafnium dioxide, tech.
Hafnium dioxide

MDL: MFCD00003565


CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: -325 Mesh Powder Density: 9.68 Odor: Odorless Merck Reference: 14,4588 MDL No.: MFCD00003565

Formula: HfO₂
MW: 210.49
Boiling Pt: 5400
Melting Pt: 2812
Density: 9.68
Storage Temperature: Ambient
MDL Number: MFCD00003565
CAS Number: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,04727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR