Aluminium oxide ≥99.9%, single crystal

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

40259-KG 40259-KJ
CAAA40259-KGEA 233.84 CAD
CAAA40259-KG CAAA40259-KJ
Aluminium oxide ≥99.9%, single crystal
Aluminium oxide

MDL: MFCD00003424


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 25.4mm dia x 3.2mm thick, random orientation MDL No.: MFCD00003424

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR