Ammonium iodide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40255-06
40255-14
40255-22
CAAA40255-06EA
118.03
CAD
CAAA40255-06
CAAA40255-14
CAAA40255-22
Ammonium iodide ≥99.995% (metals basis)
Ammonium iodide
MDL: MFCD00011424
Purity: 99.995%
CAS No.: 12027-06-4
Molecular Formula: H4IN
Formula Weight: 144.94
Physical Form: -4 Mesh
Density: 2.514
Refractive Index: 1.7031
Sensitivity: Light and Air Sensitive and Hygroscopic
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 551° subl.
Merck Reference: 14,529
MDL No.: MFCD00011424
Formula:
NH₄I Melting Pt: 551 °C (subl.) Density: 2.514 Storage Temperature: Ambient |
MDL Number:
MFCD00011424 CAS Number: 12027-06-4 EINECS: 234-717-7 Merck Index: 14,529 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...