Manganese(II) bromide tetrahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40245-14
40245-22
40245-36
CAAA40245-14EA
69.84
CAD
CAAA40245-14
CAAA40245-22
CAAA40245-36
Manganese(II) bromide tetrahydrate 98%
Manganese (II) bromide tetrahydrate
MDL: MFCD00149791
Soluble in water, alcohol
Purity: 98%
CAS No.: 10031-20-6
Molecular Formula: MnBr2.xH2O
Formula Weight: 214.76(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Pink
Odor: Odorless
Melting Point: 64.3° dec.
Solubility: Soluble in water, alcohol
Merck Reference: 14,5725
MDL No.: MFCD00149791
Formula:
Br₂Mn·4H₂O Melting Pt: 64.3 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149791 CAS Number: 10031-20-6 EINECS: 236-591-9 Merck Index: 14,5725 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...