Nickel(II) bromide trihydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
40242-18
40242-30
CAAA40242-18EA
194.34
CAD
CAAA40242-18
CAAA40242-30
AA40242-09
Nickel(II) bromide trihydrate 98%
Nickel (II) bromide trihydrate
MDL: MFCD00011141
Soluble in water, alcohol
Purity: 98%
CAS No.: 7789-49-3
Molecular Formula: NiBr2.3H2O
Formula Weight: 272.58 (218.53anhy)
Physical Form: Crystalline Granular
Sensitivity: Hygroscopic
Solubility: Soluble in water, alcohol
Merck Reference: 14,6502
UN No.: 3077
MDL No.: MFCD00011141
Formula:
Br₂Ni·3H₂O MW: 272.54 g/mol Melting Pt: 300 °C (-3H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00011141 CAS Number: 7789-49-3 EINECS: 236-665-0 UN: 3077 ADR: 9,III Merck Index: 14,6502 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...