Selenium tetrabromide 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Selenium(IV) bromide

40240-06 40240-14
CAAAA40240-06EA 95.12 CAD
CAAAA40240-06 CAAA40240-14
Selenium tetrabromide 99+%
Selenium tetrabromide

MDL: MFCD00049766 Decomposes in water. Soluble in carbon disulfide, chloroform, ethyl bromide


Purity: 99+% CAS No.: 7789-65-3 Molecular Formula: SeBr4 Formula Weight: 398.60 Physical Form: Powder Sensitivity: Moisture Sensitive Solubility: Decomposes in water. Soluble in carbon disulfide, chloroform, ethyl bromide Merck Reference: 14,8437 UN No.: 2923 MDL No.: MFCD00049766

Formula: Br₄Se
Melting Pt: 70…80 °C (dec.)
Density: 4.029 g/cm³ (–78 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049766
CAS Number: 7789-65-3
EINECS: 232-181-9
UN: 2923
ADR: 8,II
Merck Index: 14,8437

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR