Selenium chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Selenium(I) chloride
MDL: MFCD00011226
Decomposes in water. Soluble in chloroform, benzene, carbon tetrachloride, carbon disulfide
Purity: 99%
CAS No.: 10025-68-0
Molecular Formula: Se2Cl2
Formula Weight: 228.83
Physical Form: Liquid, Ampouled under argon
Density: 2.77
Melting Point: -85°
Boiling Point: 127° dec.
Solubility: Decomposes in water. Soluble in chloroform, benzene, carbon tetrachloride, carbon disulfide
Merck Reference: 14,8432
UN No.: 2922
MDL No.: MFCD00011226
Formula:
SeCl MW: 228.83 g/mol Boiling Pt: 127 °C (dec.) Melting Pt: –85 °C Density: 2.77 Storage Temperature: Ambient |
MDL Number:
MFCD00011226 CAS Number: 10025-68-0 EINECS: 233-037-8 UN: 2922 ADR: 8,II Merck Index: 14,8432 |
Specification Test Results
Form | Liquid, Ampouled under argon |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...